Anti-TIMP2 Mouse Monoclonal Antibody [clone: 89025]

Supplier: R & D Systems
MAB971
220021-728EA 772.25 USD
220021-728
Anti-TIMP2 Mouse Monoclonal Antibody [clone: 89025]
Antibodies
Type: Primary
Antigen: TIMP2
Clonality: Monoclonal
Clone: 89025
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR