Anti-CTSB Rat Monoclonal Antibody [clone: 173317]

Supplier: R & D Systems
MAB965
220021-724EA 763.57 USD
220021-724
Anti-CTSB Rat Monoclonal Antibody [clone: 173317]
Antibodies
Type: Primary
Antigen: CTSB
Clonality: Monoclonal
Clone: 173317
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR