Anti-ANGPT4 Mouse Monoclonal Antibody [clone: 15621]

Supplier: R & D Systems
MAB9641
102887-630EA 763.57 USD
102887-630
Anti-ANGPT4 Mouse Monoclonal Antibody [clone: 15621]
Antibodies
Type: Primary
Antigen: ANGPT4
Clonality: Monoclonal
Clone: 15621
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR