Anti-ANGPT4 Mouse Monoclonal Antibody [clone: 156215]

Supplier: R & D Systems
MAB964
220021-723EA 772.25 USD
220021-723
Anti-ANGPT4 Mouse Monoclonal Antibody [clone: 156215]
Antibodies
Type: Primary
Antigen: ANGPT4
Clonality: Monoclonal
Clone: 156215
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR