Anti-CLCF1 Mouse Monoclonal Antibody [clone: 138815]

Supplier: R & D Systems
MAB962
220021-722EA 772.25 USD
220021-722
Anti-CLCF1 Mouse Monoclonal Antibody [clone: 138815]
Antibodies
Type: Primary
Antigen: CLCF1
Clonality: Monoclonal
Clone: 138815
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR