Anti-DPP4 Rat Monoclonal Antibody [clone: 155202]

Supplier: R & D Systems
MAB9541
220021-716EA 763.57 USD
220021-716
Anti-DPP4 Rat Monoclonal Antibody [clone: 155202]
Antibodies
Type: Primary
Antigen: DPP4
Clonality: Monoclonal
Clone: 155202
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR