Anti-DPP4 Rat Monoclonal Antibody [clone: 155211]

Supplier: R & D Systems
MAB954
220021-715EA 772.25 USD
220021-715
Anti-DPP4 Rat Monoclonal Antibody [clone: 155211]
Antibodies
Type: Primary
Antigen: DPP4
Clonality: Monoclonal
Clone: 155211
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR