Anti-CTSL Rat Monoclonal Antibody [clone: 204106]

Supplier: R & D Systems
MAB952
220021-712EA 763.57 USD
220021-712
Anti-CTSL Rat Monoclonal Antibody [clone: 204106]
Antibodies
Type: Primary
Antigen: CTSL
Clonality: Monoclonal
Clone: 204106
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR