Anti-ADAM9 Rat Monoclonal Antibody [clone: 122823]

Supplier: R & D Systems
MAB949
220021-710EA 763.57 USD
220021-710
Anti-ADAM9 Rat Monoclonal Antibody [clone: 122823]
Antibodies
Type: Primary
Antigen: ADAM9
Clonality: Monoclonal
Clone: 122823
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR