Anti-DKK3 Rat Monoclonal Antibody [clone: 153103]

Supplier: R & D Systems
MAB948-050 MAB948-500
220021-709EA 815.63 USD
220021-709 220021-708
Anti-DKK3 Rat Monoclonal Antibody [clone: 153103]
Antibodies
Type: Primary
Antigen: DKK3
Clonality: Monoclonal
Clone: 153103
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR