Anti-DSG2 Mouse Monoclonal Antibody [clone: 141409]

Supplier: R & D Systems
MAB947
220021-707EA 763.57 USD
220021-707
Anti-DSG2 Mouse Monoclonal Antibody [clone: 141409]
Antibodies
Simple Western

Type: Primary
Antigen: DSG2
Clonality: Monoclonal
Clone: 141409
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR