Anti-ADAM15 Rat Monoclonal Antibody [clone: 1414]

Supplier: R & D Systems
MAB945
220021-705EA 772.25 USD
220021-705
Anti-ADAM15 Rat Monoclonal Antibody [clone: 1414]
Antibodies
Type: Primary
Antigen: ADAM15
Clonality: Monoclonal
Clone: 1414
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR