Anti-DSG1 Mouse Monoclonal Antibody [clone: 129204]

Supplier: R & D Systems
MAB944
220021-704EA 772.25 USD
220021-704
Anti-DSG1 Mouse Monoclonal Antibody [clone: 129204]
Antibodies
Type: Primary
Antigen: DSG1
Clonality: Monoclonal
Clone: 129204
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR