Anti-SPARC Rat Monoclonal Antibody [clone: 124413]

Supplier: R & D Systems
MAB942
220021-702EA 772.25 USD
220021-702
Anti-SPARC Rat Monoclonal Antibody [clone: 124413]
Antibodies
Type: Primary
Antigen: SPARC
Clonality: Monoclonal
Clone: 124413
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR