Anti-ADAM9 Mouse Monoclonal Antibody [clone: 138537]

Supplier: R & D Systems
MAB939
220021-700EA 772.25 USD
220021-700
Anti-ADAM9 Mouse Monoclonal Antibody [clone: 138537]
Antibodies
Type: Primary
Antigen: ADAM9
Clonality: Monoclonal
Clone: 138537
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR