Anti-TNFRSF9 Rat Monoclonal Antibody [clone: 158306]

Supplier: R & D Systems
MAB9372
220021-697EA 763.57 USD
220021-697
Anti-TNFRSF9 Rat Monoclonal Antibody [clone: 158306]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: TNFRSF9
Clonality: Monoclonal
Clone: 158306
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR