Anti-ACE2 Mouse Monoclonal Antibody [clone: 17168]

Supplier: R & D Systems
MAB9331
220021-691EA 623.29 USD
220021-691
Anti-ACE2 Mouse Monoclonal Antibody [clone: 17168]
Antibodies
Type: Primary
Antigen: ACE2
Clonality: Monoclonal
Clone: 17168
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR