Anti-ACE2 Mouse Monoclonal Antibody [clone: 17166]

Supplier: R & D Systems
MAB933
220021-690EA 763.57 USD
220021-690
Anti-ACE2 Mouse Monoclonal Antibody [clone: 17166]
Antibodies
Type: Primary
Antigen: ACE2
Clonality: Monoclonal
Clone: 17166
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR