Anti-BACE Mouse Monoclonal Antibody [clone: 137626]

Supplier: R & D Systems
MAB9311
220021-688EA 633.42 USD
220021-688
Anti-BACE Mouse Monoclonal Antibody [clone: 137626]
Antibodies
Type: Primary
Antigen: BACE
Clonality: Monoclonal
Clone: 137626
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR