Anti-ADAM17 Mouse Monoclonal Antibody [clone: 11168]

Supplier: R & D Systems
MAB9304
220021-686EA 772.25 USD
220021-686
Anti-ADAM17 Mouse Monoclonal Antibody [clone: 11168]
Antibodies
Type: Primary
Antigen: ADAM17
Clonality: Monoclonal
Clone: 11168
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR