Anti-ADAM17 Mouse Monoclonal Antibody [clone: 111636]

Supplier: R & D Systems
MAB930
220021-683EA 763.57 USD
220021-683
Anti-ADAM17 Mouse Monoclonal Antibody [clone: 111636]
Antibodies
Type: Primary
Antigen: ADAM17
Clonality: Monoclonal
Clone: 111636
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR