Anti-ACE Mouse Monoclonal Antibody [clone: 17149]

Supplier: R & D Systems
MAB9291
220021-682EA 763.57 USD
220021-682
Anti-ACE Mouse Monoclonal Antibody [clone: 17149]
Antibodies
Type: Primary
Antigen: ACE
Clonality: Monoclonal
Clone: 17149
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR