Anti-ACE Mouse Monoclonal Antibody [clone: 171417]

Supplier: R & D Systems
MAB929
220021-681EA 577.02 USD
220021-681
Anti-ACE Mouse Monoclonal Antibody [clone: 171417]
Antibodies
Type: Primary
Antigen: ACE
Clonality: Monoclonal
Clone: 171417
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR