Anti-TEK Mouse Monoclonal Antibody [clone: 176222]

Supplier: R & D Systems
MAB928
220021-680EA 763.57 USD
220021-680
Anti-TEK Mouse Monoclonal Antibody [clone: 176222]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: TEK
Clonality: Monoclonal
Clone: 176222
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR