Anti-PLG Mouse Monoclonal Antibody [clone: 79735]

Supplier: R & D Systems
MAB926
220021-679EA 763.57 USD
220021-679
Anti-PLG Mouse Monoclonal Antibody [clone: 79735]
Antibodies
Type: Primary
Antigen: PLG
Clonality: Monoclonal
Clone: 79735
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR