Anti-MMP24 Rat Monoclonal Antibody [clone: 143908]

Supplier: R & D Systems
MAB924
220021-677EA 763.57 USD
220021-677
Anti-MMP24 Rat Monoclonal Antibody [clone: 143908]
Antibodies
Type: Primary
Antigen: MMP24
Clonality: Monoclonal
Clone: 143908
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR