Anti-EDA Mouse Monoclonal Antibody [clone: 174813]

Supplier: R & D Systems
MAB922
220021-673EA 763.57 USD
220021-673
Anti-EDA Mouse Monoclonal Antibody [clone: 174813]
Antibodies
Type: Primary
Antigen: EDA
Clonality: Monoclonal
Clone: 174813
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR