Anti-MMP12 Mouse Monoclonal Antibody [clone: 82902]

Supplier: R & D Systems
MAB919
220021-672EA 772.25 USD
220021-672
Anti-MMP12 Mouse Monoclonal Antibody [clone: 82902]
Antibodies
Immunoaffinity Purification

Type: Primary
Antigen: MMP12
Clonality: Monoclonal
Clone: 82902
Conjugation: Unconjugated
Epitope: Hemopexin Domain
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR