Anti-MMP14 Mouse Monoclonal Antibody [clone: 5H2]

Supplier: R & D Systems
MAB918
220021-670EA 772.25 USD
220021-670
Anti-MMP14 Mouse Monoclonal Antibody [clone: 5H2]
Antibodies
Type: Primary
Antigen: MMP14
Clonality: Monoclonal
Clone: 5H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR