Anti-MMP15 Mouse Monoclonal Antibody [clone: 130522]

Supplier: R & D Systems
MAB9161
220021-668EA 763.57 USD
220021-668
Anti-MMP15 Mouse Monoclonal Antibody [clone: 130522]
Antibodies
Type: Primary
Antigen: MMP15
Clonality: Monoclonal
Clone: 130522
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR