Anti-MMP15 Mouse Monoclonal Antibody [clone: 130516]

Supplier: R & D Systems
MAB916
220021-667EA 772.25 USD
220021-667
Anti-MMP15 Mouse Monoclonal Antibody [clone: 130516]
Antibodies
Type: Primary
Antigen: MMP15
Clonality: Monoclonal
Clone: 130516
Conjugation: Unconjugated
Epitope: Catalytic Domain
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR