Anti-EBV IL-10 Mouse Monoclonal Antibody [clone: 134206]

Supplier: R & D Systems
MAB9151
220021-666EA 763.57 USD
220021-666
Anti-EBV IL-10 Mouse Monoclonal Antibody [clone: 134206]
Antibodies
Type: Primary
Antigen Symbol: EBV IL-10
Antigen Name:
Antigen Synonyms:
Clonality: Monoclonal
Clone: 134206
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Viral
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR