Anti-EBV IL-10 Mouse Monoclonal Antibody [clone: 134216]

Supplier: R & D Systems
MAB915
220021-665EA 763.57 USD
220021-665
Anti-EBV IL-10 Mouse Monoclonal Antibody [clone: 134216]
Antibodies
Type: Primary
Antigen Symbol: EBV IL-10
Antigen Name:
Antigen Synonyms:
Clonality: Monoclonal
Clone: 134216
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Viral
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR