Anti-ANGPTL7 Mouse Monoclonal Antibody [clone: 12148]

Supplier: R & D Systems
MAB914
220021-664EA 763.57 USD
220021-664
Anti-ANGPTL7 Mouse Monoclonal Antibody [clone: 12148]
Antibodies
Type: Primary
Antigen: ANGPTL7
Clonality: Monoclonal
Clone: 12148
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR