Anti-MMP13 Mouse Monoclonal Antibody [clone: 87518]

Supplier: R & D Systems
MAB913
220021-663EA 763.57 USD
220021-663
Anti-MMP13 Mouse Monoclonal Antibody [clone: 87518]
Antibodies
Immunoaffinity Purification

Type: Primary
Antigen: MMP13
Clonality: Monoclonal
Clone: 87518
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR