Anti-IL12 + IL23 Mouse Monoclonal Antibody [clone: 116224]

Supplier: R & D Systems
MAB9121
220021-662EA 772.25 USD
220021-662
Anti-IL12 + IL23 Mouse Monoclonal Antibody [clone: 116224]
Antibodies
Type: Primary
Antigen: IL12 + IL23
Clonality: Monoclonal
Clone: 116224
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR