Anti-IL12 + IL23 Mouse Monoclonal Antibody [clone: 116237]

Supplier: R & D Systems
MAB912
220021-661EA 763.57 USD
220021-661
Anti-IL12 + IL23 Mouse Monoclonal Antibody [clone: 116237]
Antibodies
Type: Primary
Antigen: IL12 + IL23
Clonality: Monoclonal
Clone: 116237
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR