Anti-MMP9 Mouse Monoclonal Antibody [clone: 526515]

Supplier: R & D Systems
MAB9111
220021-660EA 616.06 USD
220021-660
Anti-MMP9 Mouse Monoclonal Antibody [clone: 526515]
Antibodies
Type: Primary
Antigen: MMP9
Clonality: Monoclonal
Clone: 526515
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR