Anti-MMP9 Mouse Monoclonal Antibody [clone: 4H3]

Supplier: R & D Systems
MAB911
220021-659EA 688.37 USD
220021-659
Anti-MMP9 Mouse Monoclonal Antibody [clone: 4H3]
Antibodies
Type: Primary
Antigen: MMP9
Clonality: Monoclonal
Clone: 4H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR