Anti-MMP10 Mouse Monoclonal Antibody [clone: 110316]

Supplier: R & D Systems
MAB9102
220021-657EA 772.25 USD
220021-657
Anti-MMP10 Mouse Monoclonal Antibody [clone: 110316]
Antibodies
Type: Primary
Antigen: MMP10
Clonality: Monoclonal
Clone: 110316
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR