Anti-MMP10 Mouse Monoclonal Antibody [clone: 117239]

Supplier: R & D Systems
MAB9101
220021-656EA 763.57 USD
220021-656
Anti-MMP10 Mouse Monoclonal Antibody [clone: 117239]
Antibodies
Type: Primary
Antigen: MMP10
Clonality: Monoclonal
Clone: 117239
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR