Anti-MMP8 Mouse Monoclonal Antibody [clone: 100608]

Supplier: R & D Systems
MAB9081
220021-653EA 772.25 USD
220021-653
Anti-MMP8 Mouse Monoclonal Antibody [clone: 100608]
Antibodies
Type: Primary
Antigen: MMP8
Clonality: Monoclonal
Clone: 100608
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR