Anti-CSF2RB Mouse Monoclonal Antibody [clone: 140516]

Supplier: R & D Systems
MAB906
220021-648EA 763.57 USD
220021-648
Anti-CSF2RB Mouse Monoclonal Antibody [clone: 140516]
Antibodies
Type: Primary
Antigen: CSF2RB
Clonality: Monoclonal
Clone: 140516
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR