Anti-MMP3 Mouse Monoclonal Antibody [clone: 10D6]

Supplier: R & D Systems
MAB905
220021-647EA 633.42 USD
220021-647
Anti-MMP3 Mouse Monoclonal Antibody [clone: 10D6]
Antibodies
Type: Primary
Antigen: MMP3
Clonality: Monoclonal
Clone: 10D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR