Anti-MMP2 Mouse Monoclonal Antibody [clone: 101724]

Supplier: R & D Systems
MAB9022
220021-645EA 763.57 USD
220021-645
Anti-MMP2 Mouse Monoclonal Antibody [clone: 101724]
Antibodies
Type: Primary
Antigen: MMP2
Clonality: Monoclonal
Clone: 101724
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR