Anti-MMP2 Mouse Monoclonal Antibody [clone: 101721]

Supplier: R & D Systems
MAB9021
220021-644EA 772.25 USD
220021-644
Anti-MMP2 Mouse Monoclonal Antibody [clone: 101721]
Antibodies
Type: Primary
Antigen: MMP2
Clonality: Monoclonal
Clone: 101721
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR