Anti-MMP2 Mouse Monoclonal Antibody [clone: 36006]

Supplier: R & D Systems
MAB902
220021-643EA 647.88 USD
220021-643
Anti-MMP2 Mouse Monoclonal Antibody [clone: 36006]
Antibodies
Type: Primary
Antigen: MMP2
Clonality: Monoclonal
Clone: 36006
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR