Anti-MMP1 Mouse Monoclonal Antibody [clone: 36665]

Supplier: R & D Systems
MAB901
220021-642EA 655.11 USD
220021-642
Anti-MMP1 Mouse Monoclonal Antibody [clone: 36665]
Antibodies
Type: Primary
Antigen: MMP1
Clonality: Monoclonal
Clone: 36665
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR