Anti-CYCS Mouse Monoclonal Antibody [clone: 2B5F8]

Supplier: R & D Systems
MAB898
220021-639EA 594.37 USD
220021-639
Anti-CYCS Mouse Monoclonal Antibody [clone: 2B5F8]
Antibodies
Type: Primary
Antigen: CYCS
Clonality: Monoclonal
Clone: 2B5F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR