Anti-CYCS Mouse Monoclonal Antibody [clone: 7H8.2C12]

Supplier: R & D Systems
MAB897
220021-638EA 594.37 USD
220021-638
Anti-CYCS Mouse Monoclonal Antibody [clone: 7H8.2C12]
Antibodies
Simple Western

Type: Primary
Antigen: CYCS
Clonality: Monoclonal
Clone: 7H8.2C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR